Merck
  • Fe(3+)- and Cu(2+)-reduction by phenol derivatives associated with Azure B degradation in Fenton-like reactions.
Fe(3+)- and Cu(2+)-reduction by phenol derivatives associated with Azure B degradation in Fenton-like reactions.
Chemosphere (2006-07-15)
André Aguiar, André Ferraz
ABSTRACT
Several phenol derivatives were evaluated regarding their capacities for Fe(3+) and Cu(2+) reduction. Selected compounds were assayed in Fenton-like reactions to degrade Azure B. 3,4-Dihydroxyphenylacetic, 2,5-dihydroxyterephtalic, gallic, chromotropic and 3-hydroxyanthranilic acids were the most efficient reducers of both metallic ions. The reaction system composed of 3-hydroxyanthranilic acid/Fe(3+)/H(2)O(2) was able to degrade Azure B at higher levels than the conventional Fenton reaction (87% and 75% of decolorization after 20min reaction, respectively). Gallic and syringic acids, catechol and vanillin induced Azure B degradations at lower levels as compared with conventional Fenton reaction. Azure B was not degraded in the presence of 10% (v/v) methanol or ethanol, which are OH radical scavengers, confirming the participation of this radical in the degradation reactions. Iron-containing reactions consumed substantially more H(2)O(2) than reactions containing copper. In iron-containing reactions, even the systems that caused a limited degradation of the dye consumed high concentrations of H(2)O(2). On the other hand, the reactions containing Fe(3+), H(2)O(2) and 3-hydroxyanthranilic acid or 3,4-dihydroxyphenylacetic acid were the most efficient on degradation of Azure B and also presented the highest H(2)O(2) consumption. These results indicate that H(2)O(2) consumption occurs even when the dye is not extensively degraded, suggesting that part of the generated OH radicals reacts with the own phenol derivative instead of Azure B.
Sigma-Aldrich
Azure B, prepared by direct synthesis
Sigma-Aldrich
Azure B, certified by the Biological Stain Commission