Merck
  • Aging of mipafox-inhibited human acetylcholinesterase proceeds by displacement of both isopropylamine groups to yield a phosphate adduct.
Aging of mipafox-inhibited human acetylcholinesterase proceeds by displacement of both isopropylamine groups to yield a phosphate adduct.
Chemical research in toxicology (2006-02-21)
Timothy J Kropp, Rudy J Richardson
ABSTRACT
Aging of phosphylated serine esterases, e.g., acetylcholinesterase (AChE) and neuropathy target esterase (NTE), renders the inhibited enzymes refractory to reactivation. This process has been considered to require postinhibitory side group loss from the organophosphorus moiety. Recently, however, it has been shown that the catalytic domain of human NTE inhibited by N,N'-diisopropylphosphorodiamidofluoridate (mipafox, MIP) ages by deprotonation. For mechanistic understanding and biomarker development, it would be important to know the identity of the MIP adduct on target esterases after inhibition and aging occurred. Accordingly, the present study was performed to determine if MIP-inhibited human AChE ages by side group loss or an alternate method, e.g., deprotonation. Diisopropylphosphorofluoridate (DFP), the oxygen analogue of MIP, was used for comparison, because DFP-inhibited AChE is known to age by net loss of an isopropyl group. Kinetics experiments were done with DFP and MIP against AChE to follow the time course of inhibition, reactivation, and aging for each inhibitor. MS studies of tryptic digests from kinetically aged DFP-inhibited AChE revealed a mass shift of 122.8 +/- 0.7 Da for the active site peptide (ASP) peak, corresponding to the expected monoisopropylphosphoryl adduct. In contrast, the analogous mass shift for kinetically aged MIP-inhibited AChE was 80.7 +/- 0.9 Da, corresponding to a phosphate adduct. Because this finding was unexpected, the identity of the phosphoserine-containing ASP was confirmed by immunoprecipitation followed by MS. The results indicate that aging of MIP-inhibited AChE proceeds by displacement of both isopropylamine groups. Further research will be required to elucidate the detailed mechanism of formation of a phosphate conjugate from MIP-inhibited AChE; however, knowledge of the identity of this adduct will be useful in biomarker studies.
Sigma-Aldrich
Isopropylamine, ≥97.0% (GC)
Sigma-Aldrich
Isopropylamine, ≥99.5%
Supelco
Isopropylamine, anhydrous, analytical standard