Merck
  • Extraction capacity of diglycolamide derivatives for Ca(II), Nd(III) and Zr(IV) from nitric acid to N-dodecane containing a solvent modifier.
Extraction capacity of diglycolamide derivatives for Ca(II), Nd(III) and Zr(IV) from nitric acid to N-dodecane containing a solvent modifier.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry (2005-11-08)
Yuji Sasaki, Zhi-Xuan Zhu, Yumi Sugo, Hideya Suzuki, Takaumi Kimura
ABSTRACT
In order to evaluate the extraction property of new extractants, diglycolamide (DGA) compounds, we investigated the maximum extraction of di-, tri-, and tetravalent metal ions using nitric acid and n-dodecane. The limits of metal concentration (LOC) for Ca(II), Nd(III) and Zr(IV) in the organic phase are strongly influenced by HNO3 and the extractant concentration. For the purpose of enhancing the LOC value, we employed a modifier of the solvent, N,N-dihexyl-octanamide (DHOA) and DGA with a long alkyl chain, and examined the results. It was evident that LOC increased with the DHOA concentration and the length of the alkyl chain attached to the N atom of DGA. The stoichiometric values of LOC(Zr) estimated from the extraction reaction were confirmed by using the extraction condition: tetraoctyl-DGA/1 M DHOA + n-dodecane and 3 M HNO3.
Sigma-Aldrich
Glycolamide, 98%