Merck
Betuligenol derivative with growth inhibition and antifeedant activity.
Bioorganic & medicinal chemistry letters (2004-03-18)
Sunil K Chattopadhyay, Sachin Srivastava, Koneni V Sashidhara, Arun K Tripathi, Asish K Bhattacharya, Arvind S Negi
ABSTRACT
The title chiral amine, 3-(4-methoxyphenyl)-1-methylpropylamine 5 has been synthesized from naturally abundant betuligenol 1 in three steps and also in good yield. Furthermore, the versatile intermediate 3 could be manipulated for the preparation of chiral disulphide 7. The amine derivative 5 prepared from (-)-betuligenol showed significant growth inhibition and antifeedant activity.
Sigma-Aldrich
4-(4-Hydroxyphenyl)-2-butanone, 99%
Sigma-Aldrich
4-(4-Hydroxyphenyl)-2-butanone, ≥98%, FCC, FG
Sigma-Aldrich
4-(4-Hydroxyphenyl)-2-butanone, natural, ≥98%, FCC, FG