Merck
  • Efficient extraction of bile acid conjugates with tetraheptylammonium chloride, a liquid ion exchanger.
Efficient extraction of bile acid conjugates with tetraheptylammonium chloride, a liquid ion exchanger.
Journal of lipid research (1967-01-01)
A F Hofmann
ABSTRACT
Ethyl acetate or chloroform solutions of tetraheptylammonium chloride, an oil-soluble quaternary amine, quantitatively extract polar, anionic lipids such as steroid or bile salt conjugates from aqueous solution by a process of anion exchange.
Product Description
Supelco
Tetrahexylammonium hydrogensulfate, suitable for ion pair chromatography, LiChropur, ≥99.0% (T)
Sigma-Aldrich
Tetrahexylammonium hydrogensulfate, ≥98.0% (T)
Sigma-Aldrich
Tetrahexylammonium hydroxide solution, ~40% in H2O (T)
Supelco
Tetrahexylammonium bromide, suitable for ion pair chromatography, LiChropur, ≥99.0% (AT)
Sigma-Aldrich
Tetrahexylammonium chloride, 96%
Sigma-Aldrich
Tetrahexylammonium bromide, 99%