Merck
  • Aseptic meningitis associated with intravenous administration of dexchlorpheniramine.
Aseptic meningitis associated with intravenous administration of dexchlorpheniramine.
Annales de medecine interne (2003-08-12)
Matthieu Lafaurie, Adrien Dixmier, Jean-Michel Molina
ABSTRACT
Drug-induced aseptic meningitis has been reported mainly with the use of nonsteroidal anti-inflammatory drugs, antibiotics, intravenous immunoglobulins and OKT3 antibodies. We describe today a very unusual reaction on intravenous dexchlorpheniramine with this case of aseptic meningitis.
Product Description
Chlorphenamine maleate, European Pharmacopoeia (EP) Reference Standard
Supelco
Chlorpheniramine maleate salt solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
(±)-Chlorpheniramine maleate salt, ≥99% (perchloric acid titration)
Supelco
Chlorpheniramine Maleate, Pharmaceutical Secondary Standard; Certified Reference Material
Dexchlorpheniramine maleate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
S-(+)-Chlorpheniramine maleate salt