Merck
  • Temperature-promoted large-volume solute enrichment in column-switching miniaturized liquid chromatography: determination of an antioxidant.
Temperature-promoted large-volume solute enrichment in column-switching miniaturized liquid chromatography: determination of an antioxidant.
The Analyst (2002-08-14)
Paal Molander, Anders Holm, Elsa Lundanes, Dag R Hegna, Espen Ommundsen, Tyge Greibrokk
ABSTRACT
A two-valve sub-ambient temperature-promoted reversed-phase packed-capillary liquid-chromatography column-switching system has been tailored for sensitive determination of hydrophobic compounds. Such compounds are not easily dissolved in solvent mixtures of non-eluting properties that traditionally are used for solute enrichment in reversed-phase liquid chromatography. Enrichment-column solute focusing of large sample volumes was promoted by use of sub-ambient temperatures only, allowing the use of sample solvents that were stronger or equal to the mobile phase solvent strength. Subsequent column switching and enrichment-column temperature increment provided efficient low-dispersion back-flushed enrichment-column solute desorption onto the analytical column, where the solute was subjected to temperature-programmed gradient action. The antioxidant, Irganox 1076 (octadecyl 3-(3,5-di-tert-butyl-4-hydroxyphenyl)propionate) extracted from low density polyethylene with 100% acetonitrile served as a hydrophobic model compound. The mobile phase consisted of acetonitrile containing 10 mM triethylamine and formic acid, and the 0.25 mm id enrichment-column and analytical column in lengths of 27 and 250 mm, respectively, were packed with 3.5 microm Kromasil C18 particles. Sample volumes of up to 500 microL were successfully focused on the enrichment column at 5 degrees C using loading flow rates of up to 40 microL min(-1) prior to temperature programming to 90 degrees C. The concentration limit of detection of Irganox 1076 was 6 ng mL(-1) when using an injection volume of 500 microL. The within-assay precision was in the range 3.5-6.8% (n = 6) while the between-day precision was 7.5% (n = 3) relative standard deviation. The method was linear within the investigated mass range 3-100 ng (R2 = 0.9993).
Sigma-Aldrich
Octadecyl 3-(3,5-di-tert-butyl-4-hydroxyphenyl)propionate, 99%