Merck
  • Toward the creation of NMR databases in chiral solvents for assignments of relative and absolute stereochemistry: NMR desymmetrization of meso compounds.
Toward the creation of NMR databases in chiral solvents for assignments of relative and absolute stereochemistry: NMR desymmetrization of meso compounds.
Organic letters (2001-07-07)
Y Kobayashi, N Hayashi, Y Kishi
ABSTRACT
[structure: see text] Examples of (13)C NMR desymmetrization of meso compounds are presented. On analysis of the NMR profiles of 1,3-diols, the additivity is recognized to predict the NMR profiles of 1,3,5-tirols.
Sigma-Aldrich
N,N-Dimethylbenzylamine, ≥99%