Merck
  • In vitro cellular effects of perfluorochemicals correlate with their lipid solubility.
In vitro cellular effects of perfluorochemicals correlate with their lipid solubility.
American journal of physiology. Lung cellular and molecular physiology (2000-04-27)
V V Obraztsov, G G Neslund, E S Kornbrust, S F Flaim, C M Woods
ABSTRACT
Preclinical studies comparing perflubron partial liquid ventilation with conventional mechanical ventilation have indicated that perflubron partial liquid ventilation may exert some anti-inflammatory effects. To assess whether these effects were related to the lipid solubility properties of perflubron rather than to nonspecific biophysical properties of the perfluorocarbon (PFC) liquid phase, we studied the effects of PFCs with varying lipid solubilities on the platelet aggregation response to various procoagulants and the erythrocyte hemolytic response to osmotic stress. In both cases, the degree of the response was directly related to the lipid solubility of the PFC. All the perflubron content of erythrocytes was found to be associated with the membrane compartment. The time to reach a maximum effect on hemolysis with perflubron was relatively slow (2-4 h), which paralleled the time for perflubron to accumulate in erythrocyte membranes. The rate and extent of perflubron partitioning into lecithin liposomes were similar to those of erythrocyte membranes, supporting the hypothesis that perflubron was partitioning into the lipid component of the membranes. Thus some of the potential modulatory effects of perflubron on excessive inflammatory responses that occur during acute lung injury and acute respiratory distress syndrome may be influenced in part by the extent of PFC partitioning into the lipid bilayers of cellular membranes.
Product Description
Supelco
Heptacosafluorotributylamine, analytical standard, suitable for mass spectrometry (MS)
Sigma-Aldrich
Heptacosafluorotributylamine, liquid
Supelco
Perfluorotributylamine (PFTBA), analytical standard
Sigma-Aldrich
Perfluorohexyl bromide, 98%