Merck
  • Apoptotic cell death and impairment of L-type voltage-sensitive calcium channel activity in rat cerebellar granule cells treated with the prion protein fragment 106-126.
Apoptotic cell death and impairment of L-type voltage-sensitive calcium channel activity in rat cerebellar granule cells treated with the prion protein fragment 106-126.
Neurobiology of disease (2000-08-31)
S Thellung, T Florio, V Villa, A Corsaro, S Arena, C Amico, M Robello, M Salmona, G Forloni, O Bugiani, F Tagliavini, G Schettini
ABSTRACT
Prion diseases are neurodegenerative pathologies characterized by the accumulation, in the brain, of altered forms of the prion protein (PrP), named PrP(Sc). A synthetic peptide homologous to residues 106-126 of PrP (PrP106-126) was reported to maintain the neurodegenerative characteristics of PrP(Sc). We investigated the intracellular mechanisms involved in PrP106-126-dependent degeneration of primary cultures of cerebellar granule neurons. Prolonged exposure of such neurons to PrP106-126 induced apoptotic cell death. The L-type voltage-sensitive calcium channel blocker nicardipine reproduced this effect, suggesting that blockade of Ca(2+) entry through this class of calcium channels may be responsible for the granule cell degeneration. Microfluorometric analysis showed that PrP106-126 caused a reduction in cytosolic calcium levels, elicited by depolarizing K(+) concentrations in these neurons. Electrophysiological studies demonstrated that PrP106-126 and nicardipine selectively reduce the L-type calcium channel current. These data demonstrate that PrP106-126 alters the activity of L-type voltage-sensitive calcium channels in rat cerebellar granule cells and suggest that this phenomenon is related to the cell death induced by the peptide.
Sigma-Aldrich
Nicardipine hydrochloride, powder, ≥98%