Merck
  • Secobarbital in humans discriminating triazolam under two-response and novel-response procedures.
Secobarbital in humans discriminating triazolam under two-response and novel-response procedures.
Pharmacology, biochemistry, and behavior (1998-01-04)
J B Kamien, W K Bickel, B J Smith, G J Badger, J R Hughes
ABSTRACT
Humans were trained to discriminate the benzodiazepine triazolam (0.32 mg/70 kg) from placebo under a two-response (drug vs. placebo) drug discrimination procedure. Dose-effect curves for several drugs were then determined in a crossover design using the two-response procedure and a 'novel-response procedure' that provided a novel-appropriate response for drugs unlike triazolam or placebo. Three subjects were tested with triazolam (0.1-0.32 mg/70 kg), the barbiturate secobarbital (56-177 mg/70 kg), and caffeine (320 and 560 mg/70 kg). Triazolam dose dependently increased triazolam-appropriate responding under both procedures and generally did not occasion novel-appropriate responding under the novel-response procedure. Secobarbital substituted for triazolam in the two-response procedure and dose-dependently increased novel-appropriate responding as well as occasioned some triazolam-appropriate responding in the novel-response procedure. Caffeine generally occasioned placebo-appropriate responding under the two-response procedure and a mix of novel- and placebo-appropriate responding under the novel-response procedure. Triazolam and secobarbital produced qualitatively similar self-reported drug effects. These results suggest that the novel-response procedure for human drug discrimination may enhance the pharmacological selectivity of triazolam- and placebo-appropriate responding.
Sigma-Aldrich
Secobarbital sodium salt