Merck
  • Structural, kinetic, and pharmacodynamic mechanisms of D-amino acid oxidase inhibition by small molecules.
Structural, kinetic, and pharmacodynamic mechanisms of D-amino acid oxidase inhibition by small molecules.
Journal of medicinal chemistry (2013-05-02)
Seth C Hopkins, Michele L R Heffernan, Lakshmi D Saraswat, Carrie A Bowen, Laurence Melnick, Larry W Hardy, Michael A Orsini, Michael S Allen, Patrick Koch, Kerry L Spear, Robert J Foglesong, Mustapha Soukri, Milan Chytil, Q Kevin Fang, Steven W Jones, Mark A Varney, Aude Panatier, Stephane H R Oliet, Loredano Pollegioni, Luciano Piubelli, Gianluca Molla, Marco Nardini, Thomas H Large
ABSTRACT
We characterized the mechanism and pharmacodynamics of five structurally distinct inhibitors of d-amino acid oxidase. All inhibitors bound the oxidized form of human enzyme with affinity slightly higher than that of benzoate (Kd ≈ 2-4 μM). Stopped-flow experiments showed that pyrrole-based inhibitors possessed high affinity (Kd ≈ 100-200 nM) and slow release kinetics (k < 0.01 s(-1)) in the presence of substrate, while inhibitors with pendent aromatic groups altered conformations of the active site lid, as evidenced by X-ray crystallography, and showed slower kinetics of association. Rigid bioisosteres of benzoic acid induced a closed-lid conformation, had slower release in the presence of substrate, and were more potent than benzoate. Steady-state d-serine concentrations were described in a PK/PD model, and competition for d-serine sites on NMDA receptors was demonstrated in vivo. DAAO inhibition increased the spatiotemporal influence of glial-derived d-serine, suggesting localized effects on neuronal circuits where DAAO can exert a neuromodulatory role.
Sigma-Aldrich
Glycine, tested according to Ph. Eur.
Supelco
Glycine hydrochloride solution, 100 mM amino acid in 0.1 M HCl, analytical standard
Sigma-Aldrich
D-Amino Acid Oxidase from porcine kidney, powder, ≥1.5 units/mg solid
Supelco
Glycine, analytical standard, for nitrogen determination according to Kjeldahl method
Supelco
Glycine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Glycine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Glycine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Glycine, ACS reagent, ≥98.5%
Sigma-Aldrich
Glycine, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Glycine, BioXtra, ≥99% (titration)
Sigma-Aldrich
Glycine, 99%, FCC
SAFC
Glycine
Sigma-Aldrich
Glycine, meets analytical specification of Ph. Eur., BP, USP, 99-101% (based on anhydrous substance)
Sigma-Aldrich
Glycine, BioUltra, for molecular biology, ≥99.0% (NT)
Sigma-Aldrich
Glycine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
Sigma-Aldrich
Glycine, puriss. p.a., reag. Ph. Eur., buffer substance, 99.7-101% (calc. to the dried substance)
Sigma-Aldrich
L-Serine, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Serine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Serine, BioUltra, ≥99.5% (NT)
Supelco
L-Serine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Serine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Glycine hydrochloride, ≥99% (HPLC)
Sigma-Aldrich
Glycine 1 M solution