Merck
  • A new biomaterial of nanofibers with the microalga Spirulina as scaffolds to cultivate with stem cells for use in tissue engineering.
A new biomaterial of nanofibers with the microalga Spirulina as scaffolds to cultivate with stem cells for use in tissue engineering.
Journal of biomedical nanotechnology (2013-04-30)
D Steffens, M Lersch, A Rosa, C Scher, T Crestani, M G Morais, J A V Costa, P Pranke
ABSTRACT
The association of stem cells (SCs) with biomaterials promises to be the protagonist for future regenerative medicine in the treatment of tissue and organ lesions. Stem cells were cultivated in scaffolds constructed by the electrospinning technique, using poly-D,L-lactic acid (PDLLA) associated or not with Spirulina biomass (PDLLA/Sp), which has bioactive components of interest for tissue engineering (TE). Physicochemical analyses were performed, such as morphology, fiber diameter, degradability, residual solvent, roughness, contact angle with water, among others. SCs adhesion, proliferation and scaffold cytotoxicity were also evaluated. Nanofibers without beads and with characteristics similar to the natural extracellular matrix (ECM) in terms of mechanical and topographical properties were obtained. In biological tests it was found that SCs adhered more and had greater viability in the PDLLA/Sp molds, when compared with the PDLLA scaffolds. The scaffolds were shown to be atoxic for the SCs. It can be concluded that the scaffolds developed in this work have the characteristics to be a new biomaterial suitable for use in TE.
Sigma-Aldrich
Polylactic acid, Mw ~60,000
Sigma-Aldrich
L-Lactic Dehydrogenase from rabbit muscle, Type XI, lyophilized powder, 600-1,200 units/mg protein
Sigma-Aldrich
L-Lactic Dehydrogenase from rabbit muscle, Type II, ammonium sulfate suspension, 800-1,200 units/mg protein
Sigma-Aldrich
Lactic Dehydrogenase, recombinant from E. coli, ≥90 U/mg
Sigma-Aldrich
L-Lactic Dehydrogenase from bovine heart, 1000 units/mL
Sigma-Aldrich
L-Lactic Dehydrogenase from bovine muscle, Type X, ammonium sulfate suspension, ≥600 units/mg protein
Sigma-Aldrich
L-Lactic Dehydrogenase from bovine heart, Type XVII, buffered aqueous glycerol solution, ≥400 units/mg protein
Sigma-Aldrich
L-Lactic Dehydrogenase from porcine heart, ammonium sulfate suspension, ≥200 units/mg protein
Sigma-Aldrich
L-Lactic Dehydrogenase from bovine heart, Type III, ammonium sulfate suspension, ≥500 units/mg protein