Merck
  • Temporal changes in allocation and partitioning of new carbon as (11)C elicited by simulated herbivory suggest that roots shape aboveground responses in Arabidopsis.
Temporal changes in allocation and partitioning of new carbon as (11)C elicited by simulated herbivory suggest that roots shape aboveground responses in Arabidopsis.
Plant physiology (2013-02-02)
Abigail P Ferrieri, Beverly Agtuca, Heidi M Appel, Richard A Ferrieri, Jack C Schultz
ABSTRACT
Using the short-lived isotope (11)C (t(1/2) = 20.4 min) as (11)CO(2), we captured temporal changes in whole-plant carbon movement and partitioning of recently fixed carbon into primary and secondary metabolites in a time course (2, 6, and 24 h) following simulated herbivory with the well-known defense elicitor methyl jasmonate (MeJA) to young leaves of Arabidopsis (Arabidopsis thaliana). Both (11)CO(2) fixation and (11)C-photosynthate export from the labeled source leaf increased rapidly (2 h) following MeJA treatment relative to controls, with preferential allocation of radiolabeled resources belowground. At the same time, (11)C-photosynthate remaining in the aboveground sink tissues showed preferential allocation to MeJA-treated, young leaves, where it was incorporated into (11)C-cinnamic acid. By 24 h, resource allocation toward roots returned to control levels, while allocation to the young leaves increased. This corresponded to an increase in invertase activity and the accumulation of phenolic compounds, particularly anthocyanins, in young leaves. Induction of phenolics was suppressed in sucrose transporter mutant plants (suc2-1), indicating that this phenomenon may be controlled, in part, by phloem loading at source leaves. However, when plant roots were chilled to 5°C to disrupt carbon flow between above- and belowground tissues, source leaves failed to allocate resources belowground or toward damaged leaves following wounding and MeJA treatment to young leaves, suggesting that roots may play an integral role in controlling how plants respond defensively aboveground.
Product Description
Supelco
trans-Cinnamic acid, analytical standard
Pricing and availability is not currently available.
Sigma-Aldrich
trans-Cinnamic acid, natural, ≥99%, FCC, FG
Pricing and availability is not currently available.
Sigma-Aldrich
trans-Cinnamic acid, ≥99%
Pricing and availability is not currently available.
Sigma-Aldrich
trans-Cinnamic acid, 97%
Pricing and availability is not currently available.
Sigma-Aldrich
trans-Cinnamic acid, ≥99%, FG
Pricing and availability is not currently available.
Supelco
Invertase from baker′s yeast (S. cerevisiae), 200-300 units/mg solid
Pricing and availability is not currently available.
Sigma-Aldrich
Invertase from Candida utilis, Grade X, ≥300 units/mg solid
Pricing and availability is not currently available.
Sigma-Aldrich
Invertase from baker′s yeast (S. cerevisiae), Grade VII, ≥300 units/mg solid
Pricing and availability is not currently available.
Sigma-Aldrich
Invertase Glycoprotein Standard, BioReagent, from Saccharomyces cerevisiae
Pricing and availability is not currently available.