Merck
Targeting cancer metabolism.
Clinical cancer research : an official journal of the American Association for Cancer Research (2012-10-17)
Beverly A Teicher, W Marston Linehan, Lee J Helman
ABSTRACT
The understanding that oncogenes can have profound effects on cellular metabolism and the discovery of mutations and alterations in several metabolism-related enzymes--isocitrate dehydrogenase 1 (IDH1), isocitrate dehydrogenase 2 (IDH2), succinate dehydrogenase (SDH), fumarate hydratase (FH), and pyruvate kinase M2 (PKM2)--has renewed interest in cancer metabolism and renewed hope of taking therapeutic advantage of cancer metabolism. Otto Warburg observed that aerobic glycolysis was a characteristic of cancer cells. More than 50 years later, we understand that aerobic glycolysis and uptake of glutamine and glycine allow cancer cells to produce energy (ATP) and the nucleotides, amino acids, and lipids required for proliferation. Expression of the MYC oncogene drives the increase in cellular biomass facilitating proliferation. PKM2 expression in cancer cells stimulates aerobic glycolysis. Among intermediary metabolism enzymes, mutations in SDH occur in gastointestinal stromal tumors and result in a pseudohypoxic metabolic milieu. FH mutations lead to a characteristic renal cell carcinoma. Isocitrate dehydrogenase (IDH1/2) mutations have been found in leukemias, gliomas, prostate cancer, colon cancer, thyroid cancer, and sarcomas. These recently recognized oncogenic metabolic lesions may be selective targets for new anticancer therapeutics.
Sigma-Aldrich
Pyruvate Kinase from rabbit muscle, Type III, lyophilized powder, 350-600 units/mg protein
Sigma-Aldrich
Pyruvate Kinase from rabbit muscle, Type II, ammonium sulfate suspension, 350-600 units/mg protein
Sigma-Aldrich
Fumarase from porcine heart, ammonium sulfate suspension, ≥300 units/mg protein (biuret)
Sigma-Aldrich
Pyruvate Kinase from rabbit muscle, Type VII, buffered aqueous glycerol solution, 350-600 units/mg protein