Merck
  • An efficient and selective enzymatic oxidation system for the synthesis of enantiomerically pure D-tert-leucine.
An efficient and selective enzymatic oxidation system for the synthesis of enantiomerically pure D-tert-leucine.
Organic letters (2003-09-26)
Werner Hummel, Mutlu Kuzu, Birgit Geueke
ABSTRACT
[reaction: see text] d-tert-Leucine was prepared with an enantiomeric excess of >99% by an enzyme-catalyzed oxidative resolution of the racemic mixture of dl-tert-leucine with use of leucine dehydrogenase. The l-amino acid was oxidized completely due to coupling of the primary reaction with a highly efficient irreversible NAD(+)-regenerating step by NADH oxidase.
Sigma-Aldrich
L-Leucine Dehydrogenase from Bacillus cereus, lyophilized powder, ≥60 units/mg protein
Sigma-Aldrich
L-tert-Leucine, 99%