Merck
  • Method for differentiating Klebsiella planticola and Klebsiella terrigena from other Klebsiella species.
Method for differentiating Klebsiella planticola and Klebsiella terrigena from other Klebsiella species.
Journal of clinical microbiology (1994-04-01)
D Monnet, J Freney
ABSTRACT
Conventional methods usually fail to identify Klebsiella planticola and Klebsiella terrigena, which represent up to 19% of clinical Klebsiella isolates. By combining four carbon substrate assimilation tests and two conventional tests, the method identified these species with a specificity and a sensitivity of 100%. Overall, sensitivity for Klebsiella identification was 94.7%.
Sigma-Aldrich
D-(+)-Melezitose hydrate, ≥97% (HPLC)