Merck
Cloning of the fish cell line SSN-1 for piscine nodaviruses.
Diseases of aquatic organisms (2001-01-06)
T Iwamoto, T Nakai, K Mori, M Arimoto, I Furusawa
ABSTRACT
Six cell clones were derived from the SSN-1 cell line, which is composed of a mixed cell population and persistently infected with a C-type retrovirus (SnRV). These clones were susceptible to 4 piscine nodavirus strains belonging to different genotypes (SJNNV, RGNNV, TPNNV and *BFNNV [striped jack, redspotted grouper, tiger puffer and barfin flounder nervous necrosis viruses]). Three clones, designated A-6, E-9, and E-11, were highly permissive to nodavirus infection and production. The virus-induced cytopathic effects appeared as cytoplasmic vacuoles and intensive disintegration at 3 to 5 d post-incubation. These observations were highly reproducible and formed the basis for a successful virus titration system. Quantitative analysis using the cloned E-11 cell line clearly revealed differences in the optimal growth temperatures among the 4 genotypic variants: 25 to 30 degrees C for strain SGWak97 (RGNNV), 20 to 25 degrees C for strain SJNag93 (SJNNV), 20 degrees C for strain TPKag93 (TPNNV), and 15 to 20 degrees C for strain JFIwa98 (BFNNV). Electron microscopy demonstrated SnRV retrovirus particles only in A-6 and E-9 cells, but PCR amplification for the pol gene and LTR region of the proviral DNA indicated the presence of the retrovirus in the other clones, including E-11. The cell clones obtained in the present study will be more useful for qualitative and quantitative analyses of piscine nodaviruses than the SSN-1 cell line.
Product Description
SSN-1, NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA, 96082808, fish unknown/unspecified, Fibroblast