Merck
  • Type-2 isopentenyl diphosphate isomerase: evidence for a stepwise mechanism.
Type-2 isopentenyl diphosphate isomerase: evidence for a stepwise mechanism.
Journal of the American Chemical Society (2011-11-04)
Nicole A Heaps, C Dale Poulter
ABSTRACT
Isopentenyl diphosphate isomerase (IDI) catalyzes the interconversion of isopentenyl diphosphate (IPP) and dimethylallyl diphosphate (DMAPP). These two molecules are the building blocks for construction of isoprenoid carbon skeletons in nature. Two structurally unrelated forms of IDI are known. A variety of studies support a proton addition/proton elimination mechanism for both enzymes. During studies with Thermus thermophilus IDI-2, we discovered that the olefinic hydrogens of a vinyl thiomethyl analogue of isopentenyl diphosphate exchanged with solvent when the enzyme was incubated with D(2)O without concomitant isomerization of the double bond. These results suggest that the enzyme-catalyzed isomerization reaction is not concerted.
Sigma-Aldrich
Isopentenyl pyrophosphate trilithium salt, ≥95.0% (TLC)