Merck
  • Ex vivo Culture and Contractile Force Measurements of Non-human Primate Heart Slices.
Ex vivo Culture and Contractile Force Measurements of Non-human Primate Heart Slices.
Bio-protocol (2023-07-17)
Christine M Poch, Andreas Dendorfer, Karl-Ludwig Laugwitz, Alessandra Moretti
ABSTRACT
Cardiovascular diseases are the leading cause of death and morbidity worldwide. Patient mortality has been successfully reduced by nearly half in the last four decades, mainly due to advances in minimally invasive surgery techniques and interventional cardiology methods. However, a major hurdle is still the translational gap between preclinical findings and the conversion into effective therapies, which is partly due to the use of model systems that fail to recapitulate key aspects of human physiology and disease. Large animal models such as pigs and non-human primates are highly valuable because they closely resemble humans but are costly and time intensive. Here, we provide a method for long-term ex vivo culture of non-human primate (NHP) myocardial tissue that offers a powerful alternative for a wide range of applications including electrophysiology studies, drug screening, and gene function analyses.
Sigma-Aldrich
2,3-Butanedione monoxime, ≥98%