Merck
  • Distribution of gamma-aminobutyric acid immunoreactivity in the brain of the Siberian sturgeon (Acipenser baeri): Comparison with other fishes.
Distribution of gamma-aminobutyric acid immunoreactivity in the brain of the Siberian sturgeon (Acipenser baeri): Comparison with other fishes.
The Journal of comparative neurology (2024-02-09)
Ramón Anadón, Isabel Rodríguez-Moldes, Fátima Adrio
ABSTRACT
Gamma-aminobutyric acid (GABA) is the main inhibitory neurotransmitter in the central nervous system (CNS) of vertebrates. Immunohistochemical techniques with specific antibodies against GABA or against its synthesizing enzyme, glutamic acid decarboxylase (GAD) allowed characterizing GABAergic neurons and fibers in the CNS. However, studies on the CNS distribution of GABAergic neurons and fibers of bony fishes are scant and were done in teleost species. With the aim of understanding the early evolution of this system in bony vertebrates, we analyzed the distribution of GABA-immunoreactive (-ir) and GAD-ir neurons and fibers in the CNS of a basal ray-finned fish, the Siberian sturgeon (Chondrostei, Acipenseriformes), using immunohistochemical techniques. Our results revealed the presence and distribution of GABA/GAD-ir cells in different regions of the CNS such as olfactory bulbs, pallium and subpallium, hypothalamus, thalamus, pretectum, optic tectum, tegmentum, cerebellum, central grey, octavolateralis area, vagal lobe, rhombencephalic reticular areas, and the spinal cord. Abundant GABAergic innervation was observed in most brain regions, and GABAergic fibers were very abundant in the hypothalamic floor along the hypothalamo-hypophyseal tract and neurohypophysis. In addition, GABA-ir cerebrospinal fluid-contacting cells were observed in the alar and basal hypothalamus, saccus vasculosus, and spinal cord central canal. The distribution of GABAergic systems in the sturgeon brain shows numerous similarities to that observed in lampreys, but also to those of teleosts and tetrapods.
Sigma-Aldrich
Monoclonal Anti-GABA antibody produced in mouse, clone GB-69, ascites fluid
Sigma-Aldrich
Goat Anti-Mouse IgG Antibody, F(ab′)2, FITC conjugate, 1 mg/mL, Chemicon®