- Production and characterization of monoclonal antibodies to the catenin p120ctn.
Production and characterization of monoclonal antibodies to the catenin p120ctn.
Hybridoma (1998-06-17)
J Wu, D J Mariner, M A Thoreson, A B Reynolds
PMID9627058
ABSTRACT
This report describes the generation and characterization of a panel of monoclonal antibodies (MAb) to the catenin p120ctn. p120ctn (formerly p120cas) is a cadherin-binding protein with structural similarity to the classical catenins beta-catenin and plakoglobin. It was originally identified as a prominent Src substrate and subsequently as a substrate for the Platelet Derived Growth Factor (PDGF), Epidermal Growth Factor (EGF), and Colony Stimulating Factor-1 (CSF-1) receptor tyrosine kinases. To facilitate further study of p120 function, we have generated novel MAbs to both the N- and C-terminal ends of p120 and compared them to previously described antibodies to these regions.