Merck
  • TMEM127 suppresses tumor development by promoting RET ubiquitination, positioning, and degradation.
TMEM127 suppresses tumor development by promoting RET ubiquitination, positioning, and degradation.
Cell reports (2023-09-02)
Qianjin Guo, Zi-Ming Cheng, Hector Gonzalez-Cantú, Matthew Rotondi, Gabriela Huelgas-Morales, Purushoth Ethiraj, Zhijun Qiu, Jonathan Lefkowitz, Wan Song, Bethany N Landry, Hector Lopez, Cynthia M Estrada-Zuniga, Shivi Goyal, Mohammad Aasif Khan, Timothy J Walker, Exing Wang, Faqian Li, Yanli Ding, Lois M Mulligan, Ricardo C T Aguiar, Patricia L M Dahia
ABSTRACT
The TMEM127 gene encodes a transmembrane protein of poorly known function that is mutated in pheochromocytomas, neural crest-derived tumors of adrenomedullary cells. Here, we report that, at single-nucleus resolution, TMEM127-mutant tumors share precursor cells and transcription regulatory elements with pheochromocytomas carrying mutations of the tyrosine kinase receptor RET. Additionally, TMEM127-mutant pheochromocytomas, human cells, and mouse knockout models of TMEM127 accumulate RET and increase its signaling. TMEM127 contributes to RET cellular positioning, trafficking, and lysosome-mediated degradation. Mechanistically, TMEM127 binds to RET and recruits the NEDD4 E3 ubiquitin ligase for RET ubiquitination and degradation via TMEM127 C-terminal PxxY motifs. Lastly, increased cell proliferation and tumor burden after TMEM127 loss can be reversed by selective RET inhibitors in vitro and in vivo. Our results define TMEM127 as a component of the ubiquitin system and identify aberrant RET stabilization as a likely mechanism through which TMEM127 loss-of-function mutations cause pheochromocytoma.
Product Description
Roche
cOmplete, Mini, EDTA-free Protease Inhibitor Cocktail, Protease Inhibitor Cocktail Tablets provided in a glass vial, Tablets provided in a glass vial
Sigma-Aldrich
Leupeptin, >4000 U/mg
Sigma-Aldrich
Chloroquine diphosphate salt, powder or crystals, 98.5-101.0% (EP)
Sigma-Aldrich
Brefeldin A, ≥99% (HPLC and TLC), BioXtra, for molecular biology
Sigma-Aldrich
Bafilomycin A1
Sigma-Aldrich
Cycloheximide, ≥95% (HPLC)