Merck
  • Deterministic nuclear reprogramming of mammalian nuclei to a totipotency-like state by Amphibian meiotic oocytes for stem cell therapy in humans.
Deterministic nuclear reprogramming of mammalian nuclei to a totipotency-like state by Amphibian meiotic oocytes for stem cell therapy in humans.
Biology open (2023-11-20)
Ming-Hsuan Wen, Hector Barbosa Triana, Richard Butler, Hsiang-Wei Hu, Yang-Hong Dai, Nicola Lawrence, Jun-Jie Hong, Nigel Garrett, Rue Jones-Green, Emma L Rawlins, Ziqi Dong, Magdalena J Koziol, J B Gurdon
ABSTRACT
The ultimate aim of nuclear reprogramming is to provide stem cells or differentiated cells from unrelated cell types as a cell source for regenerative medicine. A popular route towards this is transcription factor induction, and an alternative way is an original procedure of transplanting a single somatic cell nucleus to an unfertilized egg. A third route is to transplant hundreds of cell nuclei into the germinal vesicle (GV) of a non-dividing Amphibian meiotic oocyte, which leads to the activation of silent genes in 24 h and robustly induces a totipotency-like state in almost all transplanted cells. We apply this third route for potential therapeutic use and describe a procedure by which the differentiated states of cells can be reversed so that totipotency and pluripotency gene expression are regained. Differentiated cells are exposed to GV extracts and are reprogrammed to form embryoid bodies, which shows the maintenance of stemness and could be induced to follow new directions of differentiation. We conclude that much of the reprogramming effect of eggs is already present in meiotic oocytes and does not require cell division or selection of dividing cells. Reprogrammed cells by oocytes could serve as replacements for defective adult cells in humans.
Sigma-Aldrich
DAPT, ≥98% (HPLC), solid
Sigma-Aldrich
SU-5402, ≥98% (HPLC)
Sigma-Aldrich
Smoothened Agonist, SAG, InSolution, ≥95%, 10 mM aqueous solution