Merck
  • Defining the Program of Maternal mRNA Translation during In vitro Maturation using a Single Oocyte Reporter Assay.
Defining the Program of Maternal mRNA Translation during In vitro Maturation using a Single Oocyte Reporter Assay.
Journal of visualized experiments : JoVE (2021-07-06)
Natasja G J Costermans, Enrico M Daldello, Ria J Marathe, Marco Conti
ABSTRACT
Events associated with oocyte nuclear maturation have been well described. However, much less is known about the molecular pathways and processes that take place in the cytoplasm in preparation for fertilization and acquisition of totipotency. During oocyte maturation, changes in gene expression depend exclusively on the translation and degradation of maternal messenger RNAs (mRNAs) rather than on transcription. Execution of the translational program, therefore, plays a key role in establishing oocyte developmental competence to sustain embryo development. This paper is part of a focus on defining the program of maternal mRNA translation that takes place during meiotic maturation and at the oocyte-to-zygote transition. In this method paper, a strategy is presented to study the regulation of translation of target mRNAs during in vitro oocyte maturation. Here, a Ypet reporter is fused to the 3' untranslated region (UTR) of the gene of interest and then micro-injected into oocytes together with polyadenylated mRNA encoding for mCherry to control for injected volume. By using time-lapse microscopy to measure reporter accumulation, translation rates are calculated at different transitions during oocyte meiotic maturation. Here, the protocols for oocyte isolation and injection, time-lapse recording, and data analysis have been described, using the Ypet/interleukin-7 (IL-7)-3' UTR reporter as an example.
Sigma-Aldrich
Cilostamide, A cell-permeable selective inhibitor of cGMP-inhibited phosphodiesterase (PDE III; IC₅₀ = 70 nM).