Merck
  • Colesevelam Reduces Ethanol-Induced Liver Steatosis in Humanized Gnotobiotic Mice.
Colesevelam Reduces Ethanol-Induced Liver Steatosis in Humanized Gnotobiotic Mice.
Cells (2021-07-03)
Noemí Cabré, Yi Duan, Cristina Llorente, Mary Conrad, Patrick Stern, Dennis Yamashita, Bernd Schnabl
ABSTRACT
Alcohol-related liver disease is associated with intestinal dysbiosis. Functional changes in the microbiota affect bile acid metabolism and result in elevated serum bile acids in patients with alcohol-related liver disease. The aim of this study was to identify the potential role of the bile acid sequestrant colesevelam in a humanized mouse model of ethanol-induced liver disease. We colonized germ-free (GF) C57BL/6 mice with feces from patients with alcoholic hepatitis and subjected humanized mice to the chronic-binge ethanol feeding model. Ethanol-fed gnotobiotic mice treated with colesevelam showed reduced hepatic levels of triglycerides and cholesterol, but liver injury and inflammation were not decreased as compared with non-treated mice. Colesevelam reduced hepatic cytochrome P450, family 7, subfamily a, polypeptide 1 (Cyp7a1) protein expression, although serum bile acids were not lowered. In conclusion, our findings indicate that colesevelam treatment mitigates ethanol-induced liver steatosis in mice.
Sigma-Aldrich
Colesevelam hydrochloride, ≥98% (HPLC)