Merck
  • EMSY inhibits homologous recombination repair and the interferon response, promoting lung cancer immune evasion.
EMSY inhibits homologous recombination repair and the interferon response, promoting lung cancer immune evasion.
Cell (2021-12-29)
Antonio Marzio, Emma Kurz, Jennifer M Sahni, Giuseppe Di Feo, Joseph Puccini, Shaowen Jiang, Carolina Alcantara Hirsch, Arnaldo A Arbini, Warren L Wu, Harvey I Pass, Dafna Bar-Sagi, Thales Papagiannakopoulos, Michele Pagano
ABSTRACT
Non-small cell lung cancers (NSCLCs) harboring KEAP1 mutations are often resistant to immunotherapy. Here, we show that KEAP1 targets EMSY for ubiquitin-mediated degradation to regulate homologous recombination repair (HRR) and anti-tumor immunity. Loss of KEAP1 in NSCLC induces stabilization of EMSY, producing a BRCAness phenotype, i.e., HRR defects and sensitivity to PARP inhibitors. Defective HRR contributes to a high tumor mutational burden that, in turn, is expected to prompt an innate immune response. Notably, EMSY accumulation suppresses the type I interferon response and impairs innate immune signaling, fostering cancer immune evasion. Activation of the type I interferon response in the tumor microenvironment using a STING agonist results in the engagement of innate and adaptive immune signaling and impairs the growth of KEAP1-mutant tumors. Our results suggest that targeting PARP and STING pathways, individually or in combination, represents a therapeutic strategy in NSCLC patients harboring alterations in KEAP1.
Product Description
Millipore
ANTI-FLAG® antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-EMSY antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody
Sigma-Aldrich
Puromycin dihydrochloride, Ready Made Solution, from Streptomyces alboniger, 10 mg/mL in H2O, suitable for cell culture