Merck
  • Bioluminescent and Fluorescent Reporter-Expressing Recombinant SARS-CoV-2.
Bioluminescent and Fluorescent Reporter-Expressing Recombinant SARS-CoV-2.
Methods in molecular biology (Clifton, N.J.) (2022-07-14)
Desarey Morales Vasquez, Kevin Chiem, Chengjin Ye, Luis Martinez-Sobrido
ABSTRACT
Reporter-expressing recombinant severe acute respiratory syndrome coronavirus 2 (rSARS-CoV-2) represents an excellent tool to understand the biology of and ease studying viral infections in vitro and in vivo. The broad range of applications of reporter-expressing recombinant viruses is due to the facilitated expression of fluorescence or bioluminescence readouts. In this chapter, we describe a detailed protocol on the generation of rSARS-CoV-2 expressing Venus, mCherry, and NLuc that represents a valid surrogate to track viral infections.
Sigma-Aldrich
Anti-SARS-CoV-1/2 NP Antibody, clone 1C7C7 ZooMAb® Mouse Monoclonal, recombinant, expressed in HEK 293 cells
Pricing and availability is not currently available.