Merck
  • Identification of Ebola virus microRNAs and their putative pathological function.
Identification of Ebola virus microRNAs and their putative pathological function.
Science China. Life sciences (2014-10-01)
HongWei Liang, Zhen Zhou, SuYang Zhang, Ke Zen, Xi Chen, ChenYu Zhang
ABSTRACT
Ebola virus (EBOV), a member of the filovirus family, is an enveloped negative-sense RNA virus that causes lethal infections in humans and primates. Recently, more than 1000 people have been killed by the Ebola virus disease in Africa, yet no specific treatment or diagnostic tests for EBOV are available. In this study, we identified two putative viral microRNA precursors (pre-miRNAs) and three putative mature microRNAs (miRNAs) derived from the EBOV genome. The production of the EBOV miRNAs was further validated in HEK293T cells transfected with a pcDNA6.2-GW/EmGFP-EBOV-pre-miRNA plasmid, indicating that EBOV miRNAs can be produced through the cellular miRNA processing machinery. We also predicted the potential target genes of these EBOV miRNAs and their possible biological functions. Overall, this study reports for the first time that EBOV may produce miRNAs, which could serve as non-invasive biomarkers for the diagnosis and prognosis of EBOV infection and as therapeutic targets for Ebola viral infection treatment.
Sigma-Aldrich
Anti-Dicer1 Antibody, clone 5D12.2, clone 5D12.2, Upstate®, from mouse