Merck
  • Increased vesicular γ-GABA transporter and decreased phosphorylation of synapsin I in the rostral preoptic area is associated with decreased gonadotrophin-releasing hormone and c-Fos coexpression in middle-aged female mice.
Increased vesicular γ-GABA transporter and decreased phosphorylation of synapsin I in the rostral preoptic area is associated with decreased gonadotrophin-releasing hormone and c-Fos coexpression in middle-aged female mice.
Journal of neuroendocrinology (2013-05-18)
Jing Zhang, L M Yang, X D Pan, N Lin, X C Chen
ABSTRACT
Hypothalamic glutamate (Glu) and γ-GABA neurotransmission are involved in the ovarian hormone-induced gonadotrophin-releasing hormone (GnRH)/luteinising hormone (LH) surge in rodents. Studies have shown that reduced Glu and increased γ-GABA in the rostral preoptic area (rPOA) of the hypothalamus, where most activated GnRH neurones are located, play a key role in decreasing the reproductive function of female rats. However, the mechanism underlying the altered balance of these neurotransmitters is poorly understood. In the present study, we observed a decline in the function of GnRH neurones in the rPOA at the time of the GnRH/LH surge in middle-aged intact female mice with regular oestrous cycles. In young mice, there is an increase of vesicular Glu transporter 2 on the pro-oestrus afternoon, which is not observed in middle-aged mice. By contrast, vesicular γ-GABA transporter levels in young mice decrease at the time of the LH surge, whereas they increase in middle-aged mice. Of note, we found that, in middle-aged mice at the time of the GnRH/LH surge, the phosphorylation of synapsin I at Ser603 and Ca(2+) /calmodulin-dependent kinase IIα was significantly lower than in young mice. These data suggest that, in middle-aged mice, higher levels of presynaptic stores of GABA, a lack of increase of Glu and a decreased ability of synaptic vesicle mobilisation could account for the imbalance of Glu and GABA in the rPOA, which decreases the activation of GnRH neurones.
Sigma-Aldrich
Anti-phospho-Synapsin I (pSer603) antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-GAD67 Antibody, clone 1G10.2, clone 1G10.2, Chemicon®, from mouse