Merck
  • Protein 14-3-3 Influences the Response of the Cardiac Sodium Channel Nav1.5 to Anti-Arrhythmic Drugs.
Protein 14-3-3 Influences the Response of the Cardiac Sodium Channel Nav1.5 to Anti-Arrhythmic Drugs.
The Journal of pharmacology and experimental therapeutics (2022-12-03)
Yang Zheng, Isabelle Deschenes
ABSTRACT
The cardiac sodium channel Nav1.5 is a key contributor to the cardiac action potential and dysregulations in Nav1.5 can lead to cardiac arrhythmias. Nav1.5 is a target of numerous antiarrhythmic drugs (AADs). Previous studies identified the protein 14-3-3 as a regulator of Nav1.5 biophysical coupling. Inhibition of 14-3-3 can remove the Nav1.5 functional coupling and has been shown to inhibit the dominant-negative effect of Brugada Syndrome mutations. However, it is unknown whether the coupling regulation is involved with AADs' modulation of Nav1.5. Indeed, AADs could reveal important structural and functional information about Nav1.5 coupling. Here we investigated the modulation of Nav1.5 by four classical AADs, quinidine, lidocaine, mexiletine, and flecainide, in presence of 14-3-3 inhibition. The experiments were carried out by high-throughput patch-clamp experiments in an HEK293 Nav1.5 stable cell line. We found that 14-3-3 inhibition can enhance acute block by quinidine, while the block by other drugs was not affected. We also saw changes in the use- and dose-dependency of quinidine, lidocaine, and mexiletine when inhibiting 14-3-3. Inhibiting 14-3-3 also shifted the channel activation toward hyperpolarized voltages in presence of the four drugs studied and slowed the recovery of inactivation in presence of quinidine. Our results demonstrated that the protein 14-3-3 and Nav1.5 coupling could impact the effects of AADs. Therefore, 14-3-3 and Nav1.5 coupling are new mechanisms to consider in the development of drugs targeting Nav1.5. Significance Statement The cardiac sodium channel Nav1.5 is a target of commonly used anti-arrhythmic drugs, and Nav1.5 function is regulated by the protein 14-3-3. The present study demonstrated that the regulation of Nav1.5 by 14-3-3 influences Nav1.5's response to anti-arrhythmic drugs. We provide detailed information about how 14-3-3 differentially regulated Nav1.5 functions under the influence of different drug subtypes. Our findings will guide future molecular studies investigating Nav1.5 and anti-arrhythmic drugs outcomes.
Sigma-Aldrich
Quinidine, anhydrous
Supelco
Lidocaine hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material