Merck
  • The role of SATB2 as a diagnostic marker for tumors of colorectal origin: Results of a pathology-based clinical prospective study.
The role of SATB2 as a diagnostic marker for tumors of colorectal origin: Results of a pathology-based clinical prospective study.
American journal of clinical pathology (2014-04-10)
Anca Dragomir, Meike de Wit, Christine Johansson, Mathias Uhlen, Fredrik Pontén
ABSTRACT
Immunohistochemistry is an important extension to clinical information and morphology, and prevails as an invaluable tool for establishing a correct cancer diagnosis in clinical diagnostic pathology. The applicability of immunohistochemistry is limited by the availability of validated cell- and cancer-type specific antibodies, rendering an unmet need to discover, test, and validate novel markers. The SATB2 protein is selectively expressed in glandular cells from the lower gastrointestinal tract and expression is retained in a large majority of primary and metastatic colorectal cancers. We analyzed the expression of SATB2 in all clinical cases (n = 840), in which immunohistochemistry for detection of CK20 was deemed necessary for a final diagnosis. SATB2 showed a high sensitivity (93%) and specificity (77%) to determine a cancer of colorectal origin and in combination with CK7 and CK20, the specificity increased to 100%. We conclude that SATB2 provides a new and advantageous supplement for clinical differential diagnostics.
Sigma-Aldrich
SATB2 (EP281) Rabbit Monoclonal Primary Antibody
Pricing and availability is not currently available.