Merck
  • Orexin (hypocretin) mediates light-dependent fluctuation of hippocampal function in a diurnal rodent.
Orexin (hypocretin) mediates light-dependent fluctuation of hippocampal function in a diurnal rodent.
Hippocampus (2021-07-16)
Joel E Soler, Hang Xiong, Faiez Samad, Fredric P Manfredsson, Alfred J Robison, Antonio A Núñez, Lily Yan
ABSTRACT
Environmental lighting conditions play a central role in cognitive function, but the underlying mechanisms remain unclear. Utilizing a diurnal rodent model, the Nile grass rat (Arvicanthis niloticus), we previously found that daytime light intensity affects hippocampal function in this species in a manner similar to its effects in humans. Compared to animals housed in a 12:12 h bright light-dark (brLD) cycle, grass rats kept in a 12:12 h dim light-dark (dimLD) cycle showed impaired spatial memory in the Morris water maze (MWM) and reduced CA1 apical dendritic spine density. The present study explored the neural substrates mediating the effects of daylight intensity on hippocampal function focusing on the hypothalamic orexin (hypocretin) system. First, animals housed in dimLD were treated with daily intranasal administration of orexin A peptide over five training days of the MWM task. Compared to vehicle controls, this treatment led to superior spatial memory accompanied by increased phosphorylation of Ca2+ /calmodulin-dependent protein kinase II α and glutamate receptor 1 within the CA1. To assess the role of hippocampal orexinergic signaling, an adeno-associated viral vector (AAV) expressing an orexin receptor 1 (OX1R) shRNA was injected into the dorsal hippocampus targeting the CA1 of animals housed in brLD. AAV-mediated knockdown of OX1R within the hippocampus resulted in deficits in MWM performance and reduced CA1 apical dendritic spine density. These results are consistent with the view that the hypothalamic orexinergic system underlies the modulatory role of daytime illumination on hippocampal function in diurnal mammals.
Sigma-Aldrich
Anti-CaM Kinase II Antibody, α subunit, clone 6G9, clone 6G9, Upstate®, from mouse
Sigma-Aldrich
Anti-phospho-CaM Kinase II Antibody, α subunit, (Thr286), clone 22B1, clone 22B1, Upstate®, from mouse
Sigma-Aldrich
Anti-phospho-GluR1 (Ser831) Antibody, clone N453, rabbit monoclonal, culture supernatant, clone N453, Upstate®