Merck
  • A p-Hydroxyphenacyl-Benzothiazole-Chlorambucil Conjugate as a Real-Time-Monitoring Drug-Delivery System Assisted by Excited-State Intramolecular Proton Transfer.
A p-Hydroxyphenacyl-Benzothiazole-Chlorambucil Conjugate as a Real-Time-Monitoring Drug-Delivery System Assisted by Excited-State Intramolecular Proton Transfer.
Angewandte Chemie (International ed. in English) (2016-02-27)
Shrabani Barman, Sourav K Mukhopadhyay, Sandipan Biswas, Surajit Nandi, Moumita Gangopadhyay, Satyahari Dey, Anakuthil Anoop, N D Pradeep Singh
ABSTRACT
Among the well-known phototriggers, the p-hydroxyphenacyl (pHP) group has consistently enabled the very fast, efficient, and high-conversion release of active molecules. Despite this unique behavior, the pHP group has been ignored as a delivery agent, particularly in the area of theranostics, because of two major limitations: Its excitation wavelength is below 400 nm, and it is nonfluorescent. We have overcome these limitations by incorporating a 2-(2'-hydroxyphenyl)benzothiazole (HBT) appendage capable of rapid excited-state intramolecular proton transfer (ESIPT). The ESIPT effect also provided two unique advantages: It assisted the deprotonation of the pHP group for faster release, and it was accompanied by a distinct fluorescence color change upon photorelease. In vitro studies showed that the p-hydroxyphenacyl-benzothiazole-chlorambucil conjugate presents excellent properties, such as real-time monitoring, photoregulated drug delivery, and biocompatibility.
Sigma-Aldrich
Chlorambucil