Merck
  • Nuclear localized Raf1 isoform alters DNA-dependent protein kinase activity and the DNA damage response.
Nuclear localized Raf1 isoform alters DNA-dependent protein kinase activity and the DNA damage response.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology (2018-08-15)
Benjamin R Nixon, Sara C Sebag, Michael S Glennon, Eric J Hall, Emily S Kounlavong, Michael L Freeman, Jason R Becker
ABSTRACT
Raf1/c-Raf is a well-characterized serine/threonine-protein kinase that links Ras family members with the MAPK/ERK signaling cascade. We have identified a novel splice isoform of human Raf1 that causes protein truncation and loss of the C-terminal kinase domain (Raf1-tr). We found that Raf1-tr has increased nuclear localization compared with full-length Raf1, and this finding was secondary to reduced binding of Raf1-tr to the cytoplasmic chaperone FK506 binding protein 5. We show that Raf1-tr has increased binding to DNA-dependent protein kinase (DNA-PK), which inhibits DNA-PK function and causes amplification of irradiation- and bleomycin-induced DNA damage. We found that the human colorectal cancer cell line, HCT-116, displayed reduced expression of Raf1-tr, and reintroduction of Raf1-tr sensitized the cells to bleomycin-induced apoptosis. Furthermore, we identified differential Raf1-tr expression in breast cancer cell lines and showed that breast cancer cells with increased Raf1-tr expression become sensitized to bleomycin-induced apoptosis. Collectively, these results demonstrate a novel Raf1 isoform in humans that has a unique noncanonical role in regulating the double-stranded DNA damage response pathway through modulation of DNA-PK function.-Nixon, B. R., Sebag, S. C., Glennon, M. S., Hall, E. J., Kounlavong, E. S., Freeman, M. L., Becker, J. R. Nuclear localized Raf1 isoform alters DNA-dependent protein kinase activity and the DNA damage response.
Sigma-Aldrich
RAF1 (EE) (306-end), active, GST tagged human, PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Sigma-Aldrich
Anti-Pan-Ras Antibody, clone RAS 10, clone RAS 10, from mouse
Sigma-Aldrich
Anti-Raf-1 Antibody, clone AM223, rabbit monoclonal, culture supernatant, clone AM223, from rabbit