Merck
  • Quantifying EGFR endosomal recycling via immunofluorescence in breast cancer cells.
Quantifying EGFR endosomal recycling via immunofluorescence in breast cancer cells.
STAR protocols (2022-05-03)
Ana Lonic, Winona Onglao, Yeesim Khew-Goodall
ABSTRACT
Previously published protocols for quantification of endosomal recycling are limited by the use of radioactive reagents, washing of cells in reducing buffers, or the requirement for large numbers of cells. Here, we describe a protocol for quantification of endosomal recycling using immunofluorescence that is optimized for EGFR in BT-549 breast cancer cells but could be applied to other RTKs and cell lines. Our protocol enables quick assessment of recycling and uses a relatively small number of cells. For complete details on the use and execution of this protocol, please refer to Lonic et al. (2021).
Product Description
Roche
Fibronectin, from human plasma
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, BioReagent, suitable for cell culture