Merck
  • Cap Z, a calcium insensitive capping protein in resting and activated platelets.
Cap Z, a calcium insensitive capping protein in resting and activated platelets.
FEBS letters (1996-01-15)
V T Nachmias, R Golla, J F Casella, E Barron-Casella
ABSTRACT
Capping of the barbed-ends of actin filaments is an important mechanism for control of the cytoskeleton. In platelets, a valuable model system, it has been thought that gelsolin was the major capping protein. We now report that platelets contain approximately 2 microM Cap Z, a calcium insensitive heterodimeric capping protein; two major and additional minor isoforms of both alpha and beta subunits are present. In lysates from resting platelets 75-80% of the Cap Z sediments with the high speed pellet, but if the platelets are activated with thrombin for 10 s, about 15% of the Cap Z leaves the pellet fraction and is found in the high speed supernatant where it is not bound to actin. This translocation of Cap Z to the supernatant is also observed when resting platelets are lysed into buffer containing 50-100 microM GTP gamma S and 10 mM EGTA. Our results suggest that release of Cap Z from some actin filaments could generate free filament barbed-ends. An increase in free barbed-ends has been reported in platelet lysates prepared shortly after thrombin activation.
Sigma-Aldrich
Thrombin from bovine plasma, lyophilized powder, 40-500 NIH units/mg protein (biuret)
Sigma-Aldrich
Apyrase from potatoes, ATPase ≥3.0 units/mg protein, lyophilized powder