Merck
  • Vidarabine, an anti-herpes agent, prevents occlusal-disharmony-induced cardiac dysfunction in mice.
Vidarabine, an anti-herpes agent, prevents occlusal-disharmony-induced cardiac dysfunction in mice.
The journal of physiological sciences : JPS (2022-02-13)
Yoshio Hayakawa, Kenji Suita, Yoshiki Ohnuki, Yasumasa Mototani, Misao Ishikawa, Aiko Ito, Megumi Nariyama, Akinaka Morii, Kenichi Kiyomoto, Michinori Tsunoda, Ichiro Matsuo, Hiroshi Kawahara, Satoshi Okumura
ABSTRACT
We recently reported a positive relationship between occlusal disharmony and cardiovascular disease via activation of β-adrenergic signaling in mice. Furthermore, inhibition of type 5 adenylyl cyclase (AC5), a major cardiac subtype in adults, protects the heart against oxidative stress. Here, we examined the role of AC5 in the development of occlusal-disharmony-induced cardiovascular disease in bite-opening (BO) mice, prepared by cementing a suitable appliance onto the mandibular incisor. We first examined the effects of BO treatment on cardiac function in mice treated or not treated for 2 weeks with vidarabine, which we previously identified as an inhibitor of cardiac AC. Cardiac function was significantly decreased in the BO group compared to the control group, but vidarabine ameliorated the dysfunction. Cardiac fibrosis, myocyte apoptosis and myocyte oxidative DNA damage were significantly increased in the BO group, but vidarabine blocked these changes. The BO-induced cardiac dysfunction was associated with increased phospholamban phosphorylation at threonine-17 and serine-16, as well as increased activation of the Ca2+-calmodulin-dependent protein kinase II/receptor-interacting protein 3 signaling pathway. These data suggest that AC5 inhibition with vidarabine might be a new therapeutic approach for the treatment of cardiovascular disease associated with occlusal disharmony.
Sigma-Aldrich
Anti-ADCY5 antibody produced in rabbit, affinity isolated antibody