Merck
Natural Mucosal Barriers and COVID-19 in Children.
medRxiv : the preprint server for health sciences (2021-02-18)
Carl A Pierce, Sharlene Sy, Benjamin Galen, Doctor Y Goldstein, Erika Orner, Marla Keller, Kevan C Herold, Betsy Herold
ABSTRACT
COVID-19 is more benign in children compared to adults for unknown reasons. This contrasts with viruses such as influenza where disease manifestations are often more severe in children1. We hypothesized that a more robust early innate immune response to SARS-CoV-2 may protect against severe disease and compared clinical outcomes, viral copies and cellular gene and protein expression in nasopharyngeal swabs from 12 children and 27 adults upon presentation to the Emergency Department. SARS-CoV-2 copies were similar, but compared to adults, children displayed higher expression of genes associated with interferon signaling, NLRP3 inflammasome, and other innate pathways. Higher levels of IFN-alpha2, IFN-gamma, IP-10, IL-8, and IL-1beta were detected in nasal fluid in children versus adults. Anti-SARS-CoV-2 IgA and IgG were detected in nasal fluid from both groups and correlated negatively with mucosal IL-18. These findings suggest that a more robust innate immune response in children compared to adults contributes to favorable clinical outcomes.
Product Description
Millipore
MILLIPLEX® Human Cytokine/Chemokine/Growth Factor Panel A - Immunology Multiplex Assay