Merck
  • Saturated fatty acids accelerate linear motility through mitochondrial ATP production in bull sperm.
Saturated fatty acids accelerate linear motility through mitochondrial ATP production in bull sperm.
Reproductive medicine and biology (2021-07-16)
Md Mazharul Islam, Takashi Umehara, Natsumi Tsujita, Masayuki Shimada
ABSTRACT
The present study was undertaken to clarify whether bovine sperm could take up fatty acids (FAs) and produce ATP to maintain linear motility. Frozen bovine semen was thawed in media containing either lipid mixture (LM) or FAs, and sperm motility was analyzed. The kinetic changes in FA levels in sperm were detected using gas chromatography-mass spectrometry. The mitochondrial activity of sperm thawed in media containing LM or FAs was analyzed based on the fluorescence intensity of JC-1 staining and the oxygen consumption rate. FA transporters were observed using whole-mounted immunofluorescence. Sperm linear motility was significantly (P < .05) increased after thawing in media with LM and FA. Moreover, saturated fatty acids were predominant in sperm thawed in media with LM. Notably, our study revealed that frozen bovine sperm possessed FA transporters in the midpiece where the fluorescence signals were detected after treatment with fluorescence-tagged FA. Treatment with FA activated electron transport in mitochondria through β-oxidation. Sperm linear motility is facilitated by FAs in the thawing media used for frozen bovine sperm. This might provide a new approach for upgrading the artificial insemination technique used in both livestock animals and human infertility care.
Sigma-Aldrich
Behenic acid, 99%
Sigma-Aldrich
Lipid Mixture 1, Chemically Defined, liquid, sterile-filtered, BioReagent, suitable for cell culture