Merck
  • Insulin-like growth factor binding protein-1 (IGFBP-1): a multifunctional role in the human female reproductive tract.
Insulin-like growth factor binding protein-1 (IGFBP-1): a multifunctional role in the human female reproductive tract.
Human reproduction update (2000-10-25)
D J Fowler, K H Nicolaides, J P Miell
ABSTRACT
Insulin-like growth factor-1 (IGFBP-1) is particularly important in human female reproductive physiology, where it is involved with other factors in a complex system which regulates menstrual cycles, puberty, ovulation, decidualization, implantation and fetal growth. This has implications for clinical obstetrics and gynaecology, where there is evidence for a pathophysiological role for IGFBP-1 in pre-eclampsia, intrauterine growth restriction, polycystic ovarian syndrome and trophoblast and endometrial neoplasms.