Merck
  • Identification and Structural Analysis of Spirostanol Saponin from Yucca schidigera by Integrating Silica Gel Column Chromatography and Liquid Chromatography/Mass Spectrometry Analysis.
Identification and Structural Analysis of Spirostanol Saponin from Yucca schidigera by Integrating Silica Gel Column Chromatography and Liquid Chromatography/Mass Spectrometry Analysis.
Molecules (Basel, Switzerland) (2020-08-28)
Jingya Ruan, Lu Qu, Wei Zhao, Chang Gao, Peijian Huang, Dandan Zheng, Lifeng Han, Haiyang Yu, Zixin Zhang, Yi Zhang, Tao Wang
ABSTRACT
Yucca schidigera Roezl (Mojave), a kind of ornamental plant belonging to the Yucca genus (Agavaceae), whose extract exhibits important roles in food, beverage, cosmetic and feed additives owing to its rich spirostanol saponins. To provide a comprehensive chemical profiling of the spirostanol saponins in it, this study was performed by using a multi-phase liquid chromatography method combining a reversed phase chromatography T3 column with a normal phase chromatography silica column for the separation and an ESI-Q-Exactive-Orbitrap MS in positive ion mode as the detector. By comparing the retention time and ion fragments with standards, thirty-one spirostanol saponins were identified. In addition, according to the summary of the chromatographic retention behaviors and the MS/MS cleavage patterns and biosynthetic pathway, another seventy-nine spirostanol saponins were speculatively identified, forty ones of which were potentially new ones. Moreover, ten novel spirostanol saponins (three pairs of (25R/S)-spirostanol saponin isomer mixtures) were targeted for isolation to verify the speculation. Then, the comprehensive chemical profiling of spirostanol saponins from Y. schidigera was reported here firstly.
Sigma-Aldrich
(±)-DOI hydrochloride, ≥98% (HPLC), solid