Merck
  • TGF-β1 Facilitates TAp63α Protein Lysosomal Degradation to Promote Pancreatic Cancer Cell Migration.
TGF-β1 Facilitates TAp63α Protein Lysosomal Degradation to Promote Pancreatic Cancer Cell Migration.
Biology (2021-07-03)
Guohui Gao, Jie Chen, Dongbo Wang, Qiao Li, Xiaojiao Yang, Jindan Wang, Zhiyong Pan, Zhi-Xiong Jim Xiao, Yong Yi
ABSTRACT
TGF-β signaling plays a pivotal role in promoting tumor cell migration and cancer metastasis. ΔNp63α and TAp63α are two major isoforms of p53-related p63 protein. Our recent study has shown that TGF-β1 promotes ΔNp63α protein degradation to facilitate cancer metastasis. However, whether TAp63α is involved in TGF-β1-induced cancer metastasis remains unclear. In this study, we show that, in human pancreatic cancer MIA PaCa-2 cells harboring p53-R248W allele, TGF-β1 can significantly inhibit TAp63α protein stability in a Smad pathway-independent manner. Lysosome inhibitor, chloroquine, but not proteasome inhibitor MG132, can rescue TGF-β1-induced downregulation of TAp63α protein. In addition, we show that either TGF-β1 treatment or silencing of TAp63α can dramatically increase migration of MIA PaCa-2 cells. Importantly, the restored expression of TAp63α can effectively block TGF-β1-induced migration of MIA PaCa-2 cells. Mechanistically, we show that TGF-β1 promotes TAp63α protein degradation, leading to upregulation of p53-R248W protein expression, and consequently resulting in elevated MIA PaCa-2 cell migration. Together, this study indicates that lysosomal degradation is an important way for regulating TAp63α protein fate and highlights that TGF-β1-TAp63α-mutant p53 axis is critically important in pancreatic cancer metastasis.
Sigma-Aldrich
Chloroquine diphosphate salt, powder or crystals, 98.5-101.0% (EP)
Sigma-Aldrich
TGF-β1,
Human Recombinant Animal Free
Sigma-Aldrich
(R)-MG132