Merck
  • PharmacoSTORM nanoscale pharmacology reveals cariprazine binding on Islands of Calleja granule cells.
PharmacoSTORM nanoscale pharmacology reveals cariprazine binding on Islands of Calleja granule cells.
Nature communications (2021-11-13)
Susanne Prokop, Péter Ábrányi-Balogh, Benjámin Barti, Márton Vámosi, Miklós Zöldi, László Barna, Gabriella M Urbán, András Dávid Tóth, Barna Dudok, Attila Egyed, Hui Deng, Gian Marco Leggio, László Hunyady, Mario van der Stelt, György M Keserű, István Katona
ABSTRACT
Immunolabeling and autoradiography have traditionally been applied as the methods-of-choice to visualize and collect molecular information about physiological and pathological processes. Here, we introduce PharmacoSTORM super-resolution imaging that combines the complementary advantages of these approaches and enables cell-type- and compartment-specific nanoscale molecular measurements. We exploited rational chemical design for fluorophore-tagged high-affinity receptor ligands and an enzyme inhibitor; and demonstrated broad PharmacoSTORM applicability for three protein classes and for cariprazine, a clinically approved antipsychotic and antidepressant drug. Because the neurobiological substrate of cariprazine has remained elusive, we took advantage of PharmacoSTORM to provide in vivo evidence that cariprazine predominantly binds to D3 dopamine receptors on Islands of Calleja granule cell axons but avoids dopaminergic terminals. These findings show that PharmacoSTORM helps to quantify drug-target interaction sites at the nanoscale level in a cell-type- and subcellular context-dependent manner and within complex tissue preparations. Moreover, the results highlight the underappreciated neuropsychiatric significance of the Islands of Calleja in the ventral forebrain.
Sigma-Aldrich
Glycerokinase from Cellulomonas sp., lyophilized powder, 25-75 units/mg protein
Sigma-Aldrich
Hanks′ Balanced Salt solution, Modified, with sodium bicarbonate, without phenol red, calcium chloride and magnesium sulfate, liquid, sterile-filtered, suitable for cell culture