Merck
  • Total transferrin in cerebrospinal fluid is a novel biomarker for spontaneous intracranial hypotension.
Total transferrin in cerebrospinal fluid is a novel biomarker for spontaneous intracranial hypotension.
Fukushima journal of medical science (2021-08-11)
Junko Iijima, Kyoka Hoshi, Hiromi Ito, Mayumi Kanno, Yuta Murakami, Koichi Takahashi, Kana Matsumoto, Yoshiki Yamaguchi, Madoka Nakajima, Masakazu Miyajima, Hajime Arai, Mitsuyasu Kanai, Shinobu Kitazume, Takashi Honda, Yasuhiro Hashimoto
ABSTRACT
Spontaneous intracranial hypotension (SIH) is caused by cerebrospinal fluid (CSF) leakage. Patients with SIH experience postural headaches, nausea, etc., due to CSF hypovolemia. Imaging studies and clinical examinations, such as radioisotope (RI) scintigraphy, are useful for diagnosing SIH. However, 20-30% of patients do not show typical morphology and clinical test results. We previously reported that CSF contains transferrin (Tf) isoforms:"brain-type" Tf derived from the choroid plexus and "serum-type" Tf derived from blood. We showed that both isoforms increased in the CSF of patients with SIH by Western blotting. In the present study, we demonstrate that conventional ELISA for quantifying total Tf is useful for diagnosing SIH more accurately than Western blotting. In addition, SIH with chronic subdural hematoma (CSDH) was also accurately diagnosed. Total Tf in the CSF can serve as a useful biomarker for diagnosing SIH with or without CSDH.
Sigma-Aldrich
apo-Transferrin human, powder, ≥98% (agarose gel electrophoresis)