Merck
  • Asymmetric nuclear division in neural stem cells generates sibling nuclei that differ in size, envelope composition, and chromatin organization.
Asymmetric nuclear division in neural stem cells generates sibling nuclei that differ in size, envelope composition, and chromatin organization.
Current biology : CB (2021-07-24)
Chantal Roubinet, Ian J White, Buzz Baum
ABSTRACT
Although nuclei are the defining features of eukaryotes, we still do not fully understand how the nuclear compartment is duplicated and partitioned during division. This is especially the case for organisms that do not completely disassemble their nuclear envelope upon entry into mitosis. In studying this process in Drosophila neural stem cells, which undergo asymmetric divisions, we find that the nuclear compartment boundary persists during mitosis thanks to the maintenance of a supporting nuclear lamina. This mitotic nuclear envelope is then asymmetrically remodeled and partitioned to give rise to two daughter nuclei that differ in envelope composition and exhibit a >30-fold difference in volume. The striking difference in nuclear size was found to depend on two consecutive processes: asymmetric nuclear envelope resealing at mitotic exit at sites defined by the central spindle, and differential nuclear growth that appears to depend on the available local reservoir of ER/nuclear membranes, which is asymmetrically partitioned between the two daughter cells. Importantly, these asymmetries in size and composition of the daughter nuclei, and the associated asymmetries in chromatin organization, all become apparent long before the cortical release and the nuclear import of cell fates determinants. Thus, asymmetric nuclear remodeling during stem cell divisions may contribute to the generation of cellular diversity by initiating distinct transcriptional programs in sibling nuclei that contribute to later changes in daughter cell identity and fate.
Sigma-Aldrich
Concanavalin A from Canavalia ensiformis (Jack bean), Type VI, lyophilized powder
Sigma-Aldrich
L-Glutamine solution, 200 mM, solution, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
Cytochalasin D, from Zygosporium mansonii, ≥98% (TLC and HPLC), powder
Sigma-Aldrich
Papain from papaya latex, lyophilized powder, ≥10 units/mg protein
Sigma-Aldrich
Collagenase from Clostridium histolyticum, lyophilized powder, ≥125 CDU/mg solid (CDU = collagen digestion units), 0.5-5.0 FALGPA units/mg solid
Sigma-Aldrich
Insulin from bovine pancreas, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Phalloidin, Fluorescein Isothiocyanate Labeled, sequence Amanita phalloides(synthetic: peptide sequence)
Sigma-Aldrich
Binucleine 2, ≥97% (HPLC)
Sigma-Aldrich
Latrunculin A, from sea sponge, ≥85% (HPLC), waxy solid
Sigma-Aldrich
Schneider′s Insect Medium, With L-glutamine and sodium bicarbonate, liquid, sterile-filtered, suitable for insect cell culture
Sigma-Aldrich
Cerulenin, ≥98% (HPLC), from Cephalosporium caerulens
Sigma-Aldrich
20-Hydroxyecdysone, ≥93% (HPLC), powder
Sigma-Aldrich
L-Glutathione reduced, BioXtra, ≥98.0%