Merck
  • Endothelial Tpl2 regulates vascular barrier function via JNK-mediated degradation of claudin-5 promoting neuroinflammation or tumor metastasis.
Endothelial Tpl2 regulates vascular barrier function via JNK-mediated degradation of claudin-5 promoting neuroinflammation or tumor metastasis.
Cell reports (2021-05-27)
Aikaterini Nanou, Mara Bourbouli, Stefania Vetrano, Ute Schaeper, Steven Ley, George Kollias
ABSTRACT
Increased vascular permeability and leakage are hallmarks of several pathologies and determine disease progression and severity by facilitating inflammatory/metastatic cell infiltration. Using tissue-specific genetic ablation in endothelial cells, we have investigated in vivo the role of Tumor progression locus 2 (Tpl2), a mitogen-activated protein kinase kinase kinase (MAP3K) member with pleiotropic effects in inflammation and cancer. In response to proinflammatory stimuli, endothelial Tpl2 deletion alters tight junction claudin-5 protein expression through inhibition of JNK signaling and lysosomal degradation activation, resulting in reduced vascular permeability and immune cell infiltration. This results in significantly attenuated disease scores in experimental autoimmune encephalomyelitis and fewer tumor nodules in a hematogenic lung cancer metastasis model. Accordingly, pharmacologic inhibition of Tpl2 or small interfering RNA (siRNA)-mediated Tpl2 knockdown recapitulates our findings and reduces lung metastatic tumor invasions. These results establish an endothelial-specific role for Tpl2 and highlight the therapeutic potential of blocking the endothelial-specific Tpl2 pathway in chronic inflammatory and metastatic diseases.
Sigma-Aldrich
Tpl2 Kinase Inhibitor, The Tpl2 Kinase Inhibitor, also referenced under CAS 871307-18-5, controls the biological activity of Tpl2 Kinase. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.
Sigma-Aldrich
MG-132, Ready Made Solution, ≥90% (HPLC)
Sigma-Aldrich
In Vitro Vascular Permeability Assay (24-well), This In Vitro Vascular Permeability Assay kit employs a 24-well plate and provides an efficient system for evaluating the effects of chemicals & drug compounds on endothelial cell adsorption, transport & permeability.