Merck
  • Self-organizing maps: a versatile tool for the automatic analysis of untargeted imaging datasets.
Self-organizing maps: a versatile tool for the automatic analysis of untargeted imaging datasets.
Proteomics (2013-11-26)
Pietro Franceschi, Ron Wehrens
ABSTRACT
MS-based imaging approaches allow for location-specific identification of chemical components in biological samples, opening up possibilities of much more detailed understanding of biological processes and mechanisms. Data analysis, however, is challenging, mainly because of the sheer size of such datasets. This article presents a novel approach based on self-organizing maps, extending previous work in order to be able to handle the large number of variables present in high-resolution mass spectra. The key idea is to generate prototype images, representing spatial distributions of ions, rather than prototypical mass spectra. This allows for a two-stage approach, first generating typical spatial distributions and associated m/z bins, and later analyzing the interesting bins in more detail using accurate masses. The possibilities and advantages of the new approach are illustrated on an in-house dataset of apple slices.
Sigma-Aldrich
IgG from human serum, reagent grade, ≥95% (HPLC), buffered aqueous solution
Sigma-Aldrich
IgG from human serum, reagent grade, ≥95% (SDS-PAGE), essentially salt-free, lyophilized powder