Merck
  • Diabetes-Induced H3K9 Hyperacetylation Promotes Development of Alzheimer's Disease Through CDK5.
Diabetes-Induced H3K9 Hyperacetylation Promotes Development of Alzheimer's Disease Through CDK5.
Journal of Alzheimer's disease : JAD (2020-04-21)
Hong-Bin Cai, Zhen-Zhen Fan, Ting Tian, Zi-Chao Li, Chon-Chon Zhao, Wen-Ting Guo, Zhao-Ming Ge
ABSTRACT
The connection between diabetes and Alzheimer's disease (AD) is not fully determined. Hyperphosphorylation of tau protein is mediated by binding and stabilization of truncated p25 with cyclin-dependent kinase-5 (CDK5) in AD. We recently showed that diabetes-associated hyperglycemia increased the CDK5 levels to promote development of AD. Here, we examined the underlying mechanisms. Hyperglycemia and glucose intolerance were induced in rats that had received a low dose of streptozotocin (STZ) and a high fat diet (HFD). Compared to the control rats that received no STZ and normal diet-fed, the STZ + HFD rats exhibited poorer performance in the behavioral test and showed hyperacetylation of H3K9 histone on CDK5 promoter, likely resulting from upregulation of a histone acetyltransferase, GCN5. Inhibition of acetylation of H3K9 histone by a specific GCN5 inhibitor, MB3, attenuated activation of CDK5, resulting in decreased tau phosphorylation in rat brain and improved performance of the rats in the behavior test. Thus, these data suggest that diabetes may promote future development of AD through hyperacetylation of H3K9 histone on CDK5 promoter.
Sigma-Aldrich
MB-3, ≥95% (HPLC)