Merck
  • Intermittent parathyroid hormone promotes cementogenesis via ephrinB2-EPHB4 forward signaling.
Intermittent parathyroid hormone promotes cementogenesis via ephrinB2-EPHB4 forward signaling.
Journal of cellular physiology (2020-08-03)
Tiancheng Li, Han Wang, Chunxiao Lv, Li Huang, Cheng Zhang, Chenchen Zhou, Shujuan Zou, Peipei Duan
ABSTRACT
Intermittent parathyroid hormone (PTH) promotes periodontal repair, but the underlying mechanisms remained unclear. Recent studies found that ephrinB2-EPHB4 forward signaling mediated the anabolic effect of PTH in bone homeostasis. Considering the similarities between cementum and bone, we aimed to examine the therapeutic effect of PTH on resorbed roots and explore the role of forward signaling in this process. In vivo experiments showed that intermittent PTH significantly accelerated the regeneration of root resorption and promoted expression of EPHB4 and ephrinB2. When the signaling was blocked, the resorption repair was also delayed. In vitro studies showed that intermittent PTH promoted the expression of EPHB4 and ephrinB2 in OCCM-30 cells. The effects of PTH on the mineralization capacity of OCCM-30 cells was mediated through the ephrinB2-EPHB4 forward signaling. These results support the premise that the anabolic effects of intermittent PTH on the regeneration of root resorption is via the ephrinB2-EPHB4 forward signaling pathway.
Sigma-Aldrich
Ephrin-B2/Fc Chimera from mouse, >90% (SDS-PAGE), recombinant, expressed in NSO cells, lyophilized powder
Sigma-Aldrich
Ser-Phe-Leu-Leu-Arg-Asn-Pro-Asn-Asp-Lys-Tyr-Glu-Pro-Phe, ≥97% (HPLC)
Sigma-Aldrich
NVP-BHG712, ≥98% (HPLC)